Oligo Pro II Reagents
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DN-400-0001 DN-415-0250 DN-465-1000 DN-475-1000
AGLSDN-400-0001EA 194 EUR
AGLSDN-400-0001 AGLSDN-415-0250 AGLSDN-465-1000 AGLSDN-475-1000
Oligo Pro II Reagents
Elektroforesesystemen Systemen voor capillaire elektroforese
ssDNA and ssRNA oligonucleotide purity and QC analysis.
  • High separation resolution - provides accurate assessments of oligonucleotide purity, allowing for n-1 resolution through 60-mers
  • Ready-to-use reagents - easy system setup and minimal time lost to instrument preparation, improves laboratory throughput and efficiency
  • Capillary gel electrophoresis - UV detection - No dye labeling needed
  • Allows for direct detection of both ssDNA and ssRNA
  • High throughput - load up to three 96-well plates for automated analysis
The Agilent Oligo Pro II systemMore Product Information