Safety Storage Cabinets, Type 90, V-CLASSIC-90 MULTIRISK V90.196.060.MV.2VD:0012
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40285005 40498 40627
MSGA40285005EA 6370 EUR
MSGA40285005 MSGA40498 MSGA40627
Safety Storage Cabinets, Type 90, V-CLASSIC-90 MULTIRISK V90.196.060.MV.2VD:0012
Meubilair Opslagkasten
Safe, flexible, and energetically sustainable storage of various hazardous materials in just one cabinet.
  • Special fire resistant construction
  • Locking
  • Comfortable overview and access
  • Metal-free storage levels
  • Optimal integration and use of space
With a cabinet width of only 60 cm, the MULTIRISK cabinet offers a high storage capacity (per storage level 14×1 L laboratory glass bottles or 4×2.5 L and 3×1 L laboratory glass bottles), as the cabinet with its depth of 86 cm makes perfect use of the available space next to laboratory fume cupboards. The MUTIRISK cabinet can replace three conventional safety storage cabinets, saves space in the laboratory and significantly reduces the amount of technical exhaust air required for the safe storage of hazardous materials.
Special fire resistant construction - The special, completely fire-resistant construction of the Type 90 MULTIRISK cabinet offers, for the first time, the possibility of fire-protected storage of corrosive and toxic hazardous substances. The cabinet thus not only ensures the safe storage of different hazardous materials in daily operations, but also provides maximum protection for users and rescue forces in case of a fire.
Locking - To protect against unauthorised use, the cabinet can be securely locked. The integrated red/green indicator quickly shows the locking status. The cabinet can be integrated into existing locking systems by replacing the profile cylinder on site.
Comfortable overview and access - The cabinet user is entirely unrestricted in deciding which types of hazardous materials are to be stored in the two storage compartments. Both vertical drawers can be used equally thanks to the ventilation concept and uniform interior equipment.
Metal-free storage levels - Each storage level is equipped with an tray shelf made of chemical-resistant PE and can be adjusted tool-free in height at any time by the customer. The polyethylene trays are easy to remove, e.g. for cleaning or in case of leakage.
Optimal integration and use of space - The space-saving storage solution with high storage volume is optimally suited for installation next to fume cupboards with a width of only 60 cm and a depth of 86 cm deep.
Technical ventilation - Hazardous material vapours are captured at each storage level within the cabinet and can be extracted directly via the integrated air ducts ready for connection (NW 75) to a technical exhaust system (not included). The possible formation of explosive atmospheres or harmful vapours is safely prevented.
Certificaten: Fire resistance of 90 minutes (type 90), type-tested in accordance with EN 14470-1. GS-tested. Conformity according to EN 16121 and DIN EN 16122.
Bestelinformatie: Interior equipment packages need to be ordered separately (not included). Please contact your Avantor representative for more information.
Leveringsinformatie: Delivery prices available upon request.