Guanidinium thiocyanate ≥97.0%
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
G1028-1KG G1028-20KGBL
SPCMG1028-1KGEA 1490 EUR
SPCMG1028-1KG SPCMG1028-20KGBL
Guanidinium thiocyanate ≥97.0%
Guanidinium thiocyanate
Formule: CH₅N₃·CHNS
MW: 118.16 g/mol
Smeltpunt: 118
MDL Nummer: MFCD00013027
CAS nummer: 593-84-0