N-Methylmorpholine-N-oxide 50% in aqueous solution
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR52933-250G OR52933-1KG OR52933-100G
APOSOR52933-250GEA 35.1 EUR
APOSOR52933-250G APOSOR52933-1KG APOSOR52933-100G
N-Methylmorpholine-N-oxide 50% in aqueous solution
N-Methylmorpholine-N-oxide
Formule: C₅H₁₁NO₂
Kookpunt: 119 °C
Dichtheid: 1,12
MDL Nummer: MFCD00005947
CAS nummer: 7529-22-8