5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC503021-250MG PC503021-1G PC503021-5G
APOSPC503021-250MGEA 226 EUR
APOSPC503021-250MG APOSPC503021-1G APOSPC503021-5G
5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%
5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde
Formule: C₈H₃ClF₄O
MDL Nummer: MFCD32708897
CAS nummer: 134099-32-4