5-Bromo-2-(trifluoromethoxy)phenylacetonitrile ≥98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC900121-250MG PC900121-1G
APOSPC900121-250MGEA 144 EUR
APOSPC900121-250MG APOSPC900121-1G
5-Bromo-2-(trifluoromethoxy)phenylacetonitrile ≥98%
5-Bromo-2-(trifluoromethoxy)phenylacetonitrile
Formule: C₉H₅BrF₃NO
MDL Nummer: MFCD22375096
CAS nummer: 1381944-30-4
UN: 2811