1-(4-Bromo-3,5-difluorophenyl)ethanone ≥98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC57412-1G PC57412-5G PC57412-10G
APOSPC57412-1GEA 92.1 EUR
APOSPC57412-1G APOSPC57412-5G APOSPC57412-10G
1-(4-Bromo-3,5-difluorophenyl)ethanone ≥98%
1-(4-Bromo-3,5-difluorophenyl)ethanone
Formule: C₈H₅BrF₂O
MW: 235.02 g/mol
MDL Nummer: MFCD22381231
CAS nummer: 1528607-78-4