3-Bromo-2-(bromomethyl)-1-fluoro-4-iodobenzene ≥95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC50180-250MG PC50180-1G
APOSPC50180-250MGEA 220 EUR
APOSPC50180-250MG APOSPC50180-1G
3-Bromo-2-(bromomethyl)-1-fluoro-4-iodobenzene ≥95%
3-Bromo-2-(bromomethyl)-1-fluoro-4-iodobenzene
Formule: C₇H₄Br₂FI
CAS nummer: 2092531-09-2
UN: 3261