2-Bromo-4-chloro-5-fluorobenzyl bromide 95
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC57123-250MG PC57123-1G PC57123-5G
APOSPC57123-250MGEA 191 EUR
APOSPC57123-250MG APOSPC57123-1G APOSPC57123-5G
2-Bromo-4-chloro-5-fluorobenzyl bromide 95
2-Bromo-4-chloro-5-fluorobenzyl bromide
Formule: C₇H₄Br₂ClF
MW: 302.36606 g/mol
MDL Nummer: MFCD30184439
CAS nummer: 1067882-61-4
UN: 3265