Jenway® Cuvettes
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
83070-40 83070-34 99610-33 83070-76
JENW83070-40EA 349 EUR
JENW83070-40 JENW83070-34 JENW99610-33 JENW83070-76
Jenway® Cuvettes
Cuvetten Cuvetten voor spectrofotometers
Get accurate spectroscopic readings.
  • Obtain consistent spectrophotometric data
Quartz and glass cuvettes are constructed of durable materials, making them ideal for repeated use. Single-use polystyrene cuvettes provide fast spectroscopic analysis and offer a less expensive alternative to glass and quartz cuvettes while eliminating washing and potential cross-contamination.