Reagent Reservoirs
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R225-1 R225-15 R225-3 R225-5 R225-7 R225-9
SIMPR225-1EA 499 EUR
SIMPR225-1 SIMPR225-15 SIMPR225-3 SIMPR225-5 SIMPR225-7 SIMPR225-9
Reagent Reservoirs
Reagensreservoirs
For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12 channel pipettes to 96 or 384 pipette heads.
  • Robot friendly, designed for simple, economical handling of reagents in automated systems
  • Bottom designed to enable funneling of liquid, maximising reagent recovery with pipettes
  • Low-profile
  • Chemical and heat resistant, can be stored at temperatures as low as –80 °C
  • Autoclavable
  • Can be gamma irradiated
  • Certified DNase/RNase and pyrogen free
More Product Information
Order Now
Specificaties
Environmentally Preferable