Extra Heavy-Duty Duct Tape, 389
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
389G50 389GR50 389R50 389S50 389S75
MMMA389G50EA 42.2 EUR
MMMA389G50 MMMA389GR50 MMMA389R50 MMMA389S50 MMMA389S75
Extra Heavy-Duty Duct Tape, 389
Tape
A polyethylene treated rayon cloth backing with synthetic rubber adhesive.
  • Waterproof backing has good conformability and tears easily
  • Available in multiple colours
  • Good conformability
  • Can be easily torn by hand in both directions
  • High tack adhesive and easy tear properties
Designed as a general-purpose cotton cloth tape with a high tack adhesive and easy tear properties. Used across a wide variety of applications including binding, securing, and sealing.