Secureseal™ Heat Sealing Films for Deep Well Plates
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T329-5D T329-5B
SIMPT329-5DEA 3640 EUR
SIMPT329-5D SIMPT329-5B
Secureseal™ Heat Sealing Films for Deep Well Plates
Afdichtingen Afdichtingen van microtiterplaten
These adhesive films, suppliedin sheets, are compatible with most automated adhesive SBS plate sealers.
  • Made of aluminum
  • Prevent solvent volatilization
  • No adhesive, avoiding the risk of contamination
  • Easy to peel off, time-saving and labor-saving
  • DMSO resistant
  • Can be sterilized at low temperature
Used with heat sealing machine, suitable for heat sealing deep well plates.