Bovine/Calf Blood in Heparin Sodium
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCKR500-0100EA 318 EUR
ROCKR500-0100
Bovine/Calf Blood in Heparin Sodium
Bloed
Bovine/calf blood in heparin sodium.
  • Aseptically collected and prepared to order
  • Blood draw in anticoagulant
Bovine/calf whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Bovine/calf red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.