CPC® AseptiQuik® W Genderless Aseptic Connectors
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20202-70 20202-72 20202-74 20202-76
MFLX20202-70EA 354 EUR
MFLX20202-70 MFLX20202-72 MFLX20202-74 MFLX20202-76
CPC® AseptiQuik® W Genderless Aseptic Connectors
Fittingen en connectoren
Provide easy, secure sterile connections even in nonsterile environments now in newly engineered larger sizes.
  • Genderless design simplifies system integration and eases single-use systems specifications with one part number for both halves
  • Intuitive four-step connection process reduces risk of operator error 'FLIP - CLICK - PULL - SNAP'
  • Secure side latches increase connector robustness for wide variety of applications
  • Membrane pull tabs ensure simultaneous and secure removal of both membranes
  • No secondary component needed; no need to purchase and