BRICE XXTM S3 ESD Safety Shoes, Black, Mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
760541/36 760541/37 760541/38 760541/39 760541/40 760541/41 760541/42 760541/43 760541/44 760541/45 760541/46 760541/47 760541/48
ELTN760541/36EA 90.9 EUR
ELTN760541/36 ELTN760541/37 ELTN760541/38 ELTN760541/39 ELTN760541/40 ELTN760541/41 ELTN760541/42 ELTN760541/43 ELTN760541/44 ELTN760541/45 ELTN760541/46 ELTN760541/47 ELTN760541/48
BRICE XXTM S3 ESD Safety Shoes, Black, Mid
Schoenen en laarzen
Safety shoe, BRICE XXTM, with black finish.
  • Hydrophobized microfibre material
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certificaten: EN ISO 20345 S3 SRC, form B.