jo_RAPID S1P ESD Safety Sandals, BOA® Closure
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11021/36 11021/37 11021/38 11021/39 11021/40 11021/41 11021/42 11021/43 11021/44 11021/45 11021/46 11021/47 11021/48
ELTN11021/36EA 85.9 EUR
ELTN11021/36 ELTN11021/37 ELTN11021/38 ELTN11021/39 ELTN11021/40 ELTN11021/41 ELTN11021/42 ELTN11021/43 ELTN11021/44 ELTN11021/45 ELTN11021/46 ELTN11021/47 ELTN11021/48
jo_RAPID S1P ESD Safety Sandals, BOA® Closure
Schoenen en laarzen
Safety sandal, jo_RAPID, with BOA® closure in black/red finish.
  • Microfibre material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole jo_ATHLETIC
  • Rubber scuff cap
  • Composite toe cap
For dry work areas, applications including industry, storage, transport, assembly etc. For areas where there is a risk of penetration from pointed and sharp objects (S1P) or areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certificaten: EN ISO 20345 S1P HRO/SRC, form A.