Aluminium Foil Dispensers, Korff
Leverancier: Korff
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
64900 64910 64930
293-4190EA 85.7 EUR
293-4190 293-4191 293-4193
Aluminium Foil Dispensers, Korff
Aluminiumfolie dispensers
Metallic dispensers for reels, with tear-off edge.
The core of the aluminium foil must overlap 1,5 cm on each side.