LIVAS ESD Safety Shoes, Lace-Up, Low, S1P
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720821/36 720821/37 720821/38 720821/39 720821/40 720821/41 720821/42 720821/43 720821/44 720821/45 720821/46 720821/47 720821/48 720831/36 720831/37 720831/38 720831/39 720831/40 720831/41 720831/42 720831/43 720831/44 720831/45 720831/46 720831/47 720831/48
ELTN720821/36EA 90.9 EUR
ELTN720821/36 ELTN720821/37 ELTN720821/38 ELTN720821/39 ELTN720821/40 ELTN720821/41 ELTN720821/42 ELTN720821/43 ELTN720821/44 ELTN720821/45 ELTN720821/46 ELTN720821/47 ELTN720821/48 ELTN720831/36 ELTN720831/37 ELTN720831/38 ELTN720831/39 ELTN720831/40 ELTN720831/41 ELTN720831/42 ELTN720831/43 ELTN720831/44 ELTN720831/45 ELTN720831/46 ELTN720831/47 ELTN720831/48
LIVAS ESD Safety Shoes, Lace-Up, Low, S1P
Schoenen en laarzen
ESD safety shoes, LIVAS, available in multiple color combinations.
  • Mesh material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Rubber/TPU/EVA sole TRANSFOAMERS
  • Composite toe cap
Safety shoes feature a non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S1P HRO/SRC, form A.