ESD Safety Shoes, Strap Closure, Low, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720171/39 720171/40 720171/41 720171/42 720171/43 720171/44 720171/45 720171/46 720171/47 720171/48
ELTN720171/39EA 74.9 EUR
ELTN720171/39 ELTN720171/40 ELTN720171/41 ELTN720171/42 ELTN720171/43 ELTN720171/44 ELTN720171/45 ELTN720171/46 ELTN720171/47 ELTN720171/48
ESD Safety Shoes, Strap Closure, Low, S3
Schoenen en laarzen
ESD safety shoes feature a Velcro® closure. Available in white finish.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO grey
  • MONO-PU sole SAFETY-GRIP
  • Steel toe cap
Safety shoes feature a non-metallic, penetration-resistant midsole.
Certificaten: EN ISO 20345 S3 SRC, form A.