NF-κB Inhibitor (SN50)
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17493-1 17493-5 17493-500
CAYM17493-1EA 195 EUR
CAYM17493-1 CAYM17493-5 CAYM17493-500
NF-κB Inhibitor (SN50)
Eiwitten en peptiden
A synthetic peptide corresponding to the nuclear localisation sequence of NF-κB p105 subunit (also known as p50) appended to a hydrophobic sequence to facilitate import into living cells; blocks the nuclear import of p105 in cells treated with activators of NF-κB signaling, including LPS and TNF-α.
  • A cell-permeable peptide that blocks nuclear import of NF-κB
Stability: ≥2 years
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14c33c2ebb9bd298314239f3923121c6945bf398&vid=ea7cfd90220c67407f9d7275c7ceed9f1e6ed700&prid=web&clid=SAPDAM