GenScript® GenBox Mini Electrophoresis Tank
Leverancier: Genscript
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L00780 L00781 L00782
GENSL00780EA 778 EUR
GENSL00780 GENSL00781 GENSL00782
GenScript® GenBox Mini Electrophoresis Tank
Elektroforesesystemen Gecombineerde systemen voor gel staining en blot processing
GenBox Gel electrophoresis system is designed to run 1 or 2 mini gels at the same time. GenBox mini tank is designed for ultimate convenience and consistent performance compared to traditional electrophoresis tanks. Discover the versatile and easy to use GenBox mini tank for your gel electrophoresis and blotting needs.
  • Robust: Achieve reproducible protein separation with GenBox
  • Convenience: Run up to two mini gels
  • Simple: Easy to set up and operate
  • Efficient: Compatible with Genbox mini blot module for traditional wet transfer
The GenBox mini electrophoresis tank runs both GenScript's 10*8 and 10*10 precast gels interchangeably. It can run one or two mini gels. The mini tank is compatible with GenBox blot modules for tank blotting. The GenBox Mini Trans-Blot module accommodates two gel holder cassettes, which is useful for blotting 2 mini PAGE gels at the same time.