Cole-Parmer® HT-200 Grounded Heating Tapes
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36220-10 36220-12 36220-00 36220-04 36220-06 36220-08 36220-02 36220-15 36220-14
ELET36220-10EA 927 EUR
ELET36220-10 ELET36220-12 ELET36220-00 ELET36220-04 ELET36220-06 ELET36220-08 ELET36220-02 ELET36220-15 ELET36220-14
Cole-Parmer® HT-200 Grounded Heating Tapes
Tape
Flexible heating tapes are designed to fit metal pipes and glassware.
  • Maximum element temperature of 842 °F (450 °C)
  • Braided earth wire beneath the outer sheath
  • Approximately 25 mm wide
The flexible fiberglass heating tapes are ideal for heating columns, pipes, valves, and transfer lines, where applications need to be observed. The heating element and braided ground wire are wound on a fiberglass core and covered in a braided fiberglass outer sleeve. Tapes are not water-resistant; use in dry conditions only. Tapes are supplied with 3,5'' (9 cm) insulated unheated flexible leads at each end.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.