Cole-Parmer® Essentials Replacement Cell Culturing Flasks
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21837-36 21837-35 21837-32 21837-33
TECN21837-36EA 277.04 EUR
TECN21837-36 TECN21837-35 TECN21837-32 TECN21837-33
Cole-Parmer® Essentials Replacement Cell Culturing Flasks
Kolven Celcultuurflessen
Enhance the life of your BS-200 series cell culture stirring systems.
  • Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.