Sodium chloride ≥99%, granular FCC, non-iodised
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Saline, NaCl, Sea salt, Halite
S1247-500GM S1247-12KG S1247-45KGBL
SPCMS1247-500GMEA 115 EUR
SPCMS1247-500GM SPCMS1247-12KG SPCMS1247-45KGBL
Sodium chloride ≥99%, granular FCC, non-iodised
Natriumchloride
Sodium Chloride, Non-Iodised, Granular, FCC is used as a flavor enhancer and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413 °C (1013 hPa)
Smeltpunt: 801 °C
Dichtheid: 2,16 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now
Specification Test Results
Assay 99% Min.