Potassium sulphate ≥99% FCC
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1589-500GM P1589-2.5KG P1589-12KG
SPCMP1589-500GMEA 197 EUR
SPCMP1589-500GM SPCMP1589-2.5KG SPCMP1589-12KG
Potassium sulphate ≥99% FCC
Potassium sulphate
Potassium Sulfate, FCC is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: K₂SO₄
Kookpunt: 1698 °C (1013 hPa)
Smeltpunt: 1069 °C
Dichtheid: 2,66 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011388
CAS nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759
Order Now
Specification Test Results
Assay 99% Min.