Calcium sulphate, anhydrous ≥98% FCC
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Calcium sulphate (anhydrous) anhydrite, Gypsum
CA171-2.5KG CA171-500GM
SPCMCA171-2.5KGEA 401 EUR
SPCMCA171-2.5KG SPCMCA171-500GM
Calcium sulphate, anhydrous ≥98% FCC
Calcium sulphate
Calcium Sulfate, Anhydrous, FCC is used in the food industry as an anticaking agent, dough strengthener and stabiliser. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: CaO₄S
MW: 136.14 g/mol
Smeltpunt: 1350 °C
Dichtheid: 2,964 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010912
CAS nummer: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711
Order Now
Specification Test Results
Assay 98% Min.