Chlorophyllin, coppered trisodium salt
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C3999-2.5KGBL C3999-50KGBL C3999-100GM C3999-500GM
SPCMC3999-2.5KGBLEA 4640 EUR
SPCMC3999-2.5KGBL SPCMC3999-50KGBL SPCMC3999-100GM SPCMC3999-500GM
Chlorophyllin, coppered trisodium salt
Chlorophyllin, coppered trisodium salt
Chlorophyllin Sodium Copper Salt | Non-graded Chlorophyllin Sodium Copper Salt supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formule: C₃₄H₃₁CuN₄Na₃O₆
MW: 724.15 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012149
CAS nummer: 11006-34-1