Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01189-50 01184-00 01184-02
TECN01189-50EA 7119.8 EUR
TECN01189-50 TECN01184-00 TECN01184-02
Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths
Baden Gefluïdiseerde baden
Fast heat-ups and accurate temperature control to 600 °C.
  • Dry, inert, non-toxic aluminium oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller in order to be calibrated
  • Material: Stainless steel
Use these fluidised sand baths for heat treating processes, temperature sensor calibrations and electrical component testing. Baths use aluminium oxide fluidised by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, non-flammable aluminium oxide is safer to use than oil and will not corrode sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils.
Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 57 L/min at about 3 psi pressure; order an air pump or temperature controller to accurately control the temperature of the sand bath separately. Baths feature a thermowell to hold a thermocouple thermometer.
Leveringsinformatie: Supplied with overspill flange and power cord with plug. Supplied with Type K thermocouple probe and power cord.