3-Piece Syringes
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
300188 300186 305502 308062 309649UK 309658UK 309628UK 305502UK 301231UK 303179
613-3913EA 0 EUR
613-3913 BDAM300186 BDAM305502 613-1253 BDAM309649UK BDAM309658UK 613-2997 613-2996 BDAM301231UK BDAM303179
3-Piece Syringes
Spuiten Injectiespuiten, medisch
BD Plastipak™ syringe is a precision drug delivery device that is an essential part of your daily patient care toolkit, which you can use across a range of critical applications and departments. It is intended for general purpose injection and aspiration of fluids from vials, ampoules and parts of the body below the surface of the skin.
  • 3-piece design with latex-free stopper
  • Clear barrel for visualization of syringe content
  • Bold scale printing to aid in setting for a more accurate dosage of medication
  • Special silicone lubrication ensures that plunger moves