Cole-Parmer® Stuart WS-200 Water Stills
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99295-00 99295-01
STUA99295-00EA 3260 EUR
STUA99295-00 STUA99295-01
Cole-Parmer® Stuart WS-200 Water Stills
Waterzuiveringssystemen Distilleertoestellen
Get high purity water with all-glass construction.
  • Two independent safety thermostats prevent overheating
  • Heating element shuts down when reservoir is full
This all-glass water still with silica-sheathed heaters and high-efficiency condenser ensures top distillate purity at an economical price. A reservoir level control feature automatically turns off the heater when the reservoir is full preventing any overflow. Two independent safety thermostats, mounted in the condenser and boiler, prevent overheating in the event the water supply is interrupted.
Easily and safely change the hoses without the risk of breaking any glassware using the screw thread connections. Built-in acid addition funnel and drain stopcock allow for easy descaling without dismantling the glassware. Use the still on a benchtop or wall mount using the two predrilled screw holes in the stand.
Operating pressure: 0,2 to 6,9 bar (3 to 100 psi).
Leveringsinformatie: Includes reservoir level control, predrilled support stand, hose kit, and power cord with stripped ends.