Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99294-05 99294-10 99294-20 99294-25 99294-00 99294-15
STUA99294-05EA 5840 EUR
STUA99294-05 STUA99294-10 STUA99294-20 STUA99294-25 STUA99294-00 STUA99294-15
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Waterzuiveringssystemen Distilleertoestellen
Produces cold distilled water for immediate use.
  • Provides highest purity from virtually any raw water supply
  • Reservoir water level sensor turns still off when reservoir is full
  • Simple conversion to pretreated feed
  • Safety features allow for unattended operation
  • Operating pressure: 0,2 - 6,9 bar
Remove organics, bacteria, and pyrogens from your water with the WS-200 cabinet-style water stills. Borosilicate glassware coupled with silica-sheathed heaters enclosed in a protective cabinet, deliver pyrogen-free distillate of the highest purity from virtuallyMore Product Information
Order Now
Specificaties
Model 99294-05 99294-10 99294-20 99294-25 99294-00 99294-15
Environmentally Preferable
Distillatie Single distilled Double distilled Single distilled Single distilled Single distilled Double distilled
Productie 4 L/h 4 L/h 8 L/h 8 L/h 4 L/h 4 L/h
Weerstand - bereik 0,5 - 1,0 MΩ-cm (resistivity threshold) 0,7 - 1,0 MΩ-cm (resistivity threshold) 0,5 - 1,0 MΩ-cm (resistivity threshold) 0,5 - 1,0 MΩ-cm (resistivity threshold) 0,5 - 1,0 MΩ-cm (resistivity threshold) 0,7 - 1,0 MΩ-cm (resistivity threshold)
b×d×h 550×240×410 mm 550×240×410 mm 550×240×410 mm 550×240×410 mm 550×240×410 mm 550×240×410 mm