Printer Ribbons, M710 R6900 Series
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173183 173181 173182 173184 173180
BRDY173183EA 76.22 EUR
BRDY173183 BRDY173181 BRDY173182 BRDY173184 BRDY173180
Printer Ribbons, M710 R6900 Series
Printerlinten
One-color ribbon for printing a single color onto a label.
  • Recommended for use on glossy or smooth label and sign materials
  • When printed on the appropriate label material, it has excellent solvent, scratch, and fade resistance
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.