Inkjet Polypropylene Roll Labels Brady® BradyJet™ 76,2 mm Core
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153646 153647
BRDY153646EA 261.7 EUR
BRDY153646 BRDY153647
Inkjet Polypropylene Roll Labels Brady® BradyJet™ 76,2 mm Core
Etiketten
B-2569 inkjet-printable polyester tape from Brady® features a specially-formulated topcoat for improved durability and colour.
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol, and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Designed for roll inkjet printers
  • Matte finish
Applications include electrical labelling, facility identification, general identification, hazard communication identification, lean and 5S labelling, pipe and valve marking, safety identification, and warehouse marking.