Core Inkjet Repositionable Vinyl Roll Labels
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153648 153649 153650
BRDY153648EA 255.64 EUR
BRDY153648 BRDY153649 BRDY153650
Core Inkjet Repositionable Vinyl Roll Labels
Etiketten
Thanks to a special aggressive adhesive, B-2581 inkjet-printable vinyl labels from Brady® stay in place when needed, and remove cleanly without leaving residue.
  • Durable and flexible material makes it ideal for areas where identification is frequently changed
  • Designed for roll inkjet printers
  • Service temperature of −40 to +60 °C (−40 to 140 °F)
  • Matte finish
Applications include general identification, lean and 5S labelling, safety identification, and temporary labelling.