Core Inkjet Water Dissolvable Paper Roll Labels
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153641 153657
BRDY153641EA 81.29 EUR
BRDY153641 BRDY153657
Core Inkjet Water Dissolvable Paper Roll Labels
Etiketten
This B-403 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.
  • Ideal for use in laboratory settings, asset tracking, or general use
  • An easy to remove, environmentally responsible, print-on-demand inkjet label solution
  • Easy removal saves time during relabeling process
  • Pressure-sensitive adhesive allows for quick and easy application
  • Designed for roll inkjet printers
  • Matte finish
Applications include asset tracking, general identification, and laboratory labelling.