Inkjet Polypropylene Roll Labels Brady® BradyJet™ 50,80 mm Core
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153637 153638 153640
BRDY153637EA 165.09 EUR
BRDY153637 BRDY153638 BRDY153640
Inkjet Polypropylene Roll Labels Brady® BradyJet™ 50,80 mm Core
Etiketten
This B-7425J polypropylene material is ideal for full-colour identification and barcode applications where resistance to liquid nitrogen, autoclave conditions, and harsh chemicals is required.
  • White polypropylene with acrylic adhesive is inkjet printable
  • Perfect for laboratory identification where labels may come in contact with harsh chemicals
  • Designed for roll inkjet printers
  • Service temperature of −80 to +70 °C (−112 to +158 °F)
  • Matte finish
Applications include barcode labelling, cold temperature labelling, COVID-19, general identification, and laboratory labelling.