NovaSnap™ Mops
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MZNS6-14 MZNS6-14IR
MICRMZNS6-14EA 467 EUR
MICRMZNS6-14 MICRMZNS6-14IR
NovaSnap™ Mops
Moppen
This low-touch flat mop uses a simple snap-on, step-release clip action for improved cleanliness and ergonomics. Simply align the components and push into place. The ergonomic unit weighs less than 2 pounds (0,8 kilos) – handle, adapter, and NovaSnap™ mop.
  • Cover snaps securely onto mop frame
  • Step-on release system reduces contact
  • Well suited for use on equipment, floors, walls, and windows
  • Evenly coats surfaces with disinfectants
  • Equally effective for damp mopping
  • Urethane foam interiors fully encased in 100% smooth polyester
  • Suitable for use with strong disinfectants and repeated autoclaving
  • Recyclable
The Micronova NovaSnap™ Mop Covers snap securely onto the 360° swivel SlimLine™ adapter, which can be locked at 180° for more controlled cleaning. Available gamma irradiated for aseptic environments.
Bestelinformatie: All components are ordered separately. For additional handle lengths, see STAEH series plastic mop handles.
Verpakking: Individually bagged.