PermaSleeve® Heat Shrink Wire and Cable Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174582 174652 174578
BRDY174582EA 108.62 EUR
BRDY174582 BRDY174652 BRDY174578
PermaSleeve® Heat Shrink Wire and Cable Labels for M710 and BMP71
Etiketten
PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use and permanence to create long-lasting, high-quality identification.
  • 3:1 heat shrinkable sleeve fit snugly around wires for maximum insulation protection and permanent identification
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • Cold resistant and flame-retardant with an operating temperature of −55 to +135 °C (−67 to +275 °F)
  • Matte finish
Applications include electrical labelling and wire and cable labelling.
A ribbon is required to print this material. Use part number M7-R4300.
Certificaten: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202. RoHS compliant. UL recognised.