DuraSleeve® Wire Marking Inserts for M611, M610 and M710
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312051 312050 312062 312061 312098 312097
BRDY312051EA 43.65 EUR
BRDY312051 BRDY312050 BRDY312062 BRDY312061 BRDY312098 BRDY312097
DuraSleeve® Wire Marking Inserts for M611, M610 and M710
Etiketten
The DuraSleeve® wire marking system provides printable inserts that can easily be inserted into a clean carrier.
  • DuraSleeve® markers for the M610 and M611 printers
  • DuraSleeve® markers create a durable legend
  • The legend can be changed before or after termination
  • Matte finish
Applications include wire and cable labelling.
Certificaten: Halogen-free (DIN VDE 0472/part 815).
Verpakking: 1500 inserts per box.