Powder Funnels, PP
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0016600 0016700 0016800 0016900 0017000 0017100
221-1536EA 47.2 EUR
221-1536 KART0016700 KART0016800 KART0016900 KART0017000 KART0017100
Powder Funnels, PP
Trechters Poedertrechters
60° angle, autoclavable funnels.
  • Large bore avoids bridging of powders during transfer
  • Ribbed outside to avoid pressure lock
  • Excellent chemical resistance
Can be used with liquids.
Certificaten: The raw materials used comply with food-contact EU regulation 1935/2004, 10/2011, US Food and Drug Administration and REACH/ROHS regulation.