Overturnable Stoppers, Silicone Rubber
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0119100 0119300 0119400 0119500 0119600 0119700 0119000 0119200
KART0119100EA 94.7 EUR
KART0119100 KART0119300 KART0119400 KART0119500 KART0119600 KART0119700 KART0119000 KART0119200
Overturnable Stoppers, Silicone Rubber
Stoppen Siliconen stoppen
Pierceable stoppers to withdraw solutions and suspensions from closed containers, using syringes.
Suitable for use in food and beverage industry.
Height (H/h): H: height of overturnable section; h: height of lower section.