Test Tubes with Screw Caps
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8830404 8830406 8830410
KART8830404EA 421 EUR
KART8830404 KART8830406 KART8830410
Test Tubes with Screw Caps
Buisjes Reageerbuisjes
Test tubes with quick closure screw caps (PE).
  • Leak-proof
  • Suitable for centrifugation
  • Available in blue, red and yellow
The design of the cap avoids any contamination due to drops of liquid on the inner surface. Supplied with the screw cap fitted on the test tube.