Shodex HILICpak VC-50 HILIC Columns for Cationic Analytes Including Amines
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F6711600 F7630700
SHOWF6711600EA 676 EUR
SHOWF6711600 SHOWF7630700
Shodex HILICpak VC-50 HILIC Columns for Cationic Analytes Including Amines
Kolommen voor chromatografie
Robust polymer-based HILIC columns for positively charged analytes.
  • Modified carboxyl group is suitable for cationic substance analysis including amines
  • Best for aminoglycoside antibiotics, histamine/histidine, monoamine neurotransmitters and more
  • The dominant separation mode is RP or IEX rather than HILIC mode
  • Polymer-based packing material provides excellent chemical stability and minimum deterioration over an extended time period
  • pH stability: 2 to 13 (silanol free)
  • Easily regenerated by washing in an alkaline solution
  • Also suitable for evaporative