Amersham ECL Direct Labeling and Detection System, Cytiva
Leverancier: CytivaTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 minute chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.
- Direct probe labelling in a 10 minute reaction, 1 hour from hybridization to detection with ECL Direct and Hybond™ N+.
- Eliminates handling, waste and regulatory issues associated with use of radioactivity.
- No need to strip blots before reprobing.
- Contains a novel rate enhancer, giving higher sensitivity with very low backgrounds on nylon membranes.
Leveringsinformatie: ECL Direct is particularly useful for applications that require a blot to be reprobed several times and also for screening applications requiring rapid throughput and fast results. Examples including High-to-Medium-target Southern and Northen blots, Colony/ Plaque lifts, Dot or Slot Blots, Genome Mapping, PCR Product Analysis, Library Screens.