b.safe Fittings PFAS Analytic
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M401-06 M401-18
BOHLM401-06EA 45.9 EUR
BOHLM401-06 BOHLM401-18
b.safe Fittings PFAS Analytic
Fittingen en connectoren
Fittings made of PE to fix tubing in the capillary connectors (thread UNF ¹/₄" 28G) of b.safe Caps PFAS Analytic.
The conical seal cone clamps the tubing and seals the connection safely.