CE & CE/MS Supplies, Agilent Technologies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5063-6514 9300-1747 G1600-67201 G1607-60041
HEWL5063-6514EA 333 EUR
HEWL5063-6514 HEWL9300-1747 HEWLG1600-67201 HEWLG1607-60041
CE & CE/MS Supplies, Agilent Technologies
Elektroforesesystemen Systemen voor capillaire elektroforese
CE/MS sprayer kit and CE/MS adapter kit to couple an Agilent CE instrument with an MS system.
Agilent provides CE and CE/MS supplies, including sprayer kits and CE adapter kits, to couple an Agilent CE instrument with MS systems.The CE/MS sprayer kit needs the CE/MS adapter kit to fully support CE/MS coupling.