Cole-Parmer® Stuart V Vortex Mixers
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
04729-03 04729-01 04729-07 04729-13
STUA04729-03EA 613 EUR
STUA04729-03 STUA04729-01 STUA04729-07 STUA04729-13
Cole-Parmer® Stuart V Vortex Mixers
Schudapparaten Vortexmengers
Compact unit perfect for individual work stations.
  • Mix a wide variety of tube sizes and shapes with the included rubber cup head
  • Heavy die-cast body won't dance across the bench
  • Automatic start when the soft rubber cup is pressed down
The vortex mixer provides rapid mixing of samples contained in test-tubes, small flasks, and bottles. Starts automatically when the rubber cup is depressed and stops once vessel is removed. Heavy die-cast body for stability during use.
The fixed speed Stuart Vortex Mixer measures 7.8×7.8×21.5 cm (W×H×D) with a 4.2 mm orbit size. The variable speed Stuart Vortex Mixer measures 7.8×7.8×21.5 cm (W×H×D) with a 4.2 mm orbit size. The variable speed Stuart Mini Vortex Mixer measures 10×6.7×10 cm (W×H×D) with a 4.5 mm orbit size.
Leveringsinformatie: All Stuart Vortex Mixers include standard rubber cup. The Accessory Cup Pack is designed for use with variable speed Vortex Mixer 04729-03, and includes (1) plastic cradle for standard microtitre plate, (1) circular foam insert for beakers and flasks (up to 500 ml capacity), (1) rectangular foam insert (holds 8×0.2, 8×0.5 and 16×1.5 ml microcentrifuge tubes), and (1) solid rectangular foam insert (blank for custom holes).