BASE Footwear Grand Canyon Safety Ankle Shoes, Lace-UP, Mid, S1P
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0667BKR35 B0667BKR36 B0667BKR37 B0667BKR38 B0667BKR39 B0667BKR40 B0667BKR41 B0667BKR42 B0667BKR43 B0667BKR44 B0667BKR45 B0667BKR46 B0667BKR47 B0667BKR48
PRTWB0667BKR35EA 147 EUR
PRTWB0667BKR35 PRTWB0667BKR36 PRTWB0667BKR37 PRTWB0667BKR38 PRTWB0667BKR39 PRTWB0667BKR40 PRTWB0667BKR41 PRTWB0667BKR42 PRTWB0667BKR43 PRTWB0667BKR44 PRTWB0667BKR45 PRTWB0667BKR46 PRTWB0667BKR47 PRTWB0667BKR48
BASE Footwear Grand Canyon Safety Ankle Shoes, Lace-UP, Mid, S1P
Schoenen en laarzen
Grand Canyon mid safety shoes feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
  • 200 joules resistant toe-cap
  • Puncture-resistant sole
  • Shock absorption in the heel region
  • Anti-static footwear
  • Hydrocarbons-resistant
  • Electronic devices protection from electrostatic discharges
  • Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Certificaten: EN ISO 20345: 2011 S1P FO ESD SR; CEI EN 61340-4-3 S1P.