BASE Footwear Yellowstone Safety Shoes, Lace-Up, Low, S3
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0666BBKR35 B0666BBKR36 B0666BBKR37 B0666BBKR38 B0666BBKR39 B0666BBKR40 B0666BBKR41 B0666BBKR42 B0666BBKR43 B0666BBKR44 B0666BBKR45 B0666BBKR46 B0666BBKR47 B0666BBKR48
PRTWB0666BBKR35EA 139 EUR
PRTWB0666BBKR35 PRTWB0666BBKR36 PRTWB0666BBKR37 PRTWB0666BBKR38 PRTWB0666BBKR39 PRTWB0666BBKR40 PRTWB0666BBKR41 PRTWB0666BBKR42 PRTWB0666BBKR43 PRTWB0666BBKR44 PRTWB0666BBKR45 PRTWB0666BBKR46 PRTWB0666BBKR47 PRTWB0666BBKR48
BASE Footwear Yellowstone Safety Shoes, Lace-Up, Low, S3
Schoenen en laarzen
Yellowstone low safety shoes feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
  • 200 joules resistant toe-cap
  • Puncture-resistant sole
  • Shock absorption in the heel region
  • Anti-static footwear
  • Electronic devices protection from electrostatic discharges
  • Hydrocarbons-resistant
  • Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Certificaten: More Product Information
Order Now
Specificaties
Environmentally Preferable